SY3H315B119C Synthonix Corporation

5-Methyl-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD03011381
  • Cas Number : 3757-53-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/36732.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/36732.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart