SY3H315B11A4 Synthonix Corporation

3-Methyl-1,2,4-triazine

  • MDL number MFCD10000820
  • Cas Number : 24108-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/36785.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/36785.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart