SY3H315B11A6 Synthonix Corporation

6-Methoxy-3(2H)-benzofuranone

  • MDL number MFCD02257681
  • Cas Number : 15832-09-4
  • Molecular Weight : 164.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-methoxy-1-benzofuran-3(2H)-one
Inchi Code 1S/C9H8O3/c1-11-6-2-3-7-8(10)5-12-9(7)4-6/h2-5,10H,1H3
Inchi Key DNBWDCJLHVOQHX-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/36796.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/36796.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart