SY3H315B11A9 Synthonix Corporation

(2-(Methylthio)pyrimidin-5-yl)methanol

  • MDL number MFCD11053756
  • Cas Number : 19858-50-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to Pale-yellow Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/36801.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/36801.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart