SY3H315B11B3 Synthonix Corporation

Methyl 5-(bromomethyl)picolinate

  • MDL number MFCD07367952
  • Cas Number : 55876-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to off-white to pale-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/36849.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/36849.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart