SY3H315B11B4 Synthonix Corporation

Methyl 2-oxo-1,2-dihydropyrimidine-4-carboxylate

  • MDL number MFCD21608027
  • Cas Number : 950514-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Off-white to light yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/36852.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/36852.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart