SY3H315B11D8 Synthonix Corporation

Methyl 1H-pyrrolo[2,3-c]pyridine-3-carboxylate

  • MDL number MFCD16036494
  • Cas Number : 108128-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37117.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37117.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart