SY3H315B11F0 Synthonix Corporation

Methyl 2-amino-1H-benzo[d]imidazole-5-carboxylate

  • MDL number MFCD08752985
  • Cas Number : 106429-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37270.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37270.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart