SY3H315B121C Synthonix Corporation

(2-Methyloxazol-5-yl)methanol

  • MDL number MFCD11101415
  • Cas Number : 1065073-48-4
  • Molecular Weight : 113.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-methyl-1,3-oxazol-5-yl)methanol
Inchi Code 1S/C5H7NO2/c1-4-6-2-5(3-7)8-4/h2,7H,3H2,1H3
Inchi Key RLZPINFKUMSNAK-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/37561.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37561.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart