SY3H315B1224 Synthonix Corporation

Methyl 2-amino-4-bromo-5-fluorobenzoate

  • MDL number No MDL found
  • Cas Number : 1395493-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/37617.html

Safety Information

MSDS https://synthonix.com/37617.html
Availability details for
Quantity
  • Availability
  • Add to Cart