SY3H315B1246 Synthonix Corporation

6-Nitropyridin-2-amine

  • MDL number MFCD04114202
  • Cas Number : 14916-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Pale-yellow to orange to yellow-brown solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/36745.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/36745.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart