SY3H315B125C Synthonix Corporation

5H-Pyrido[4,3-b]indole

  • MDL number MFCD00965976
  • Cas Number : 244-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Pale reddish yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/36832.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/36832.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart