SY3H315B1278 Synthonix Corporation

1-Phenyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

  • MDL number MFCD16659786
  • Cas Number : 1002334-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37731.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37731.html
pictograms GHS07
hstatements H302, H315, H319, H332, H335
Availability details for
Quantity
  • Availability
  • Add to Cart