SY3H315B12CD Synthonix Corporation

2-(1H-1,2,4-Triazol-3-yl)pyridine

  • MDL number MFCD13188589
  • Cas Number : 23195-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Slightly pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37759.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37759.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart