SY3H315B12CF Synthonix Corporation

2-Vinylaniline

  • MDL number MFCD11878483
  • Cas Number : 3867-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/36733.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/36733.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart