SY3H315B12D2 Synthonix Corporation

2-Vinylphenylboronic acid

  • MDL number MFCD01074659
  • Cas Number : 15016-42-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/5638.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/5638.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart