SY3H315B12D6 Synthonix Corporation

(2-Oxo-2,3-dihydrobenzo[d]oxazol-6-yl)boronic acid

  • MDL number MFCD18072745
  • Cas Number : 1016644-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/32436.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/32436.html
pictograms GHS07
hstatements H302, H315, H319, H332, H335
Availability details for
Quantity
  • Availability
  • Add to Cart