SY3H315B12DF Synthonix Corporation

(R)-tert-Butyl (2-oxooxetan-3-yl)carbamate

  • MDL number MFCD11519129
  • Cas Number : 126330-77-6
  • Molecular Weight : 187.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (3R)-2-oxo-3-oxetanylcarbamate
Inchi Code 1S/C8H13NO4/c1-8(2,3)13-7(11)9-5-4-12-6(5)10/h5H,4H2,1-3H3,(H,9,11)/t5-/m1/s1
Inchi Key HRJDEHQWXAPGBG-RXMQYKEDSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form White to Yellow Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/37079.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37079.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart