SY3H315B12E1 Synthonix Corporation

(S)-Oxiran-2-ylmethyl 3-nitrobenzenesulfonate

  • MDL number MFCD00064582
  • Cas Number : 115314-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37186.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37186.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart