SY3H3249F009 Synthonix Corporation

C-Azetidin-2-yl-methylamine dihydrochloride

  • MDL number MFCD22415246
  • Cas Number : 1389264-23-6
  • Molecular Weight : 159.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7337.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/7337.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart