SY3H3249F00A Synthonix Corporation

4-tert-Butyldimethylsiloxy-1-butanol

  • MDL number MFCD04974070
  • Cas Number : 87184-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/0335.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/0335.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart