SY3H3249F011 Synthonix Corporation

3-Aminotetrahydrofuran-3-carboxylic acid

  • MDL number MFCD08460976
  • Cas Number : 125218-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/0696.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/0696.html
hstatements H315 , H317 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart