SY3H3249F014 Synthonix Corporation

4-(Aminomethyl)-1-methyl-2-pyrrolidinone

  • MDL number No MDL found
  • Cas Number : 933723-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/10326.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/10326.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart