SY3H3249F028 Synthonix Corporation

3-Amino-1-Boc-4,4-difluoropyrrolidine

  • Cas Number : 1408074-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/11641.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/11641.html
hstatements H302 , H315 , H318 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart