SY3H3249F045 Synthonix Corporation

3-(Boc-amino)tetrahydrofuran-3-methanol

  • MDL number MFCD23106437
  • Cas Number : 1132814-98-2
  • Molecular Weight : 217.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/13187.html

Safety Information

signalword Warning
pcodes P202, P261, P280
MSDS https://synthonix.com/13187.html
pictograms GHS07
hstatements H302, H320
Availability details for
Quantity
  • Availability
  • Add to Cart