SY3H3249F05D Synthonix Corporation

3-Aminomethyl-3-fluoropiperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD18375048
  • Cas Number : 1219832-36-6
  • Molecular Weight : 232.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form oil
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/1464.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/1464.html
pictograms GHS05, GHS07
hstatements H302, H315, H318, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart