SY3H3249F060 Synthonix Corporation

3-(Boc-aminomethyl)-3-methylpyrrolidine

  • MDL number No MDL found
  • Cas Number : 125290-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/14769.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/14769.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart