SY3H3249F085 Synthonix Corporation

tert-butyl N-{2-azaspiro[3.4]octan-6-yl}carbamate

  • MDL number No MDL found
  • Cas Number : 1638761-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/21121.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/21121.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart