SY3H3249F0A0 Synthonix Corporation

4-Amino-1-(tetrahydropyranyl)-1H-indazole

  • MDL number MFCD10699168
  • Cas Number : 1053655-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/3177.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/3177.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart