SY3H3249F0A2 Synthonix Corporation

6-Amino-1-(tetrahydropyranyl)-1H-indazole

  • MDL number MFCD10699171
  • Cas Number : 1053655-59-6
  • Molecular Weight : 217.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/3180.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/3180.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart