SY3H3249F0C2 Synthonix Corporation

3-Amino-2-fluorotoluene

  • MDL number MFCD06410915
  • Cas Number : 1978-33-2
  • Molecular Weight : 125.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Clear liquid / Colour: Colorless - Red - Brown
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/37863.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37863.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart