SY3H3249F0CC Synthonix Corporation

tert-Butyl (4-aminocyclohexyl)carbamate

  • MDL number No MDL found
  • Cas Number : 195314-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/37949.html

Safety Information

MSDS https://synthonix.com/37949.html
Availability details for
Quantity
  • Availability
  • Add to Cart