SY3H3249F13B Synthonix Corporation

N-Butoxycarbonyl trimethylphosphonoglycine

  • MDL number MFCD02178893
  • Cas Number : 89524-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/0033.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/0033.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart