SY3H3249F13D Synthonix Corporation

1-Amino-7-azaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester hydrochloride

  • MDL number MFCD19439666
  • Cas Number : 1100748-78-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/6055.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6055.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart