SY3H3249F142 Synthonix Corporation

2-Amino-6-tert-butyl-4,5,6,7-tetrahydrobenzothiazole

  • MDL number MFCD02854748
  • Cas Number : 81779-11-5
  • Molecular Weight : 210.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/6982.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/6982.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart