SY3H3249F147 Synthonix Corporation

6-Amino-1,4,8-trioxaspiro[4.5]decane oxalate

  • MDL number MFCD22573370
  • Cas Number : 1392803-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7283.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7283.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart