SY3H3249F15B Synthonix Corporation

1-Benzyl-2-methyl-piperidin-4-one

  • MDL number MFCD00480711
  • Cas Number : 203661-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/0125.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/0125.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart