SY3H3249F15C Synthonix Corporation

Tert-butyl 4-hydroxyazepane-1-carboxylate

  • MDL number MFCD07368610
  • Cas Number : 478832-21-2
  • Molecular Weight : 215.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/0130.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/0130.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart