SY3H3249F160 Synthonix Corporation

1-(tert-Butoxycarbonyl)-3-iodopyrrolidine

  • MDL number MFCD12031283
  • Cas Number : 774234-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/0289.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/0289.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart