SY3H3249F165 Synthonix Corporation

3-Bromotetrahydrofuran

  • MDL number MFCD09056777
  • Cas Number : 19311-37-6
  • Molecular Weight : 151

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Freezer
Physical Form Liquid
Shipping Temperature Ice Pak
Country of Origin US
Product Link https://synthonix.com/0393.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/0393.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart