SY3H3249F166 Synthonix Corporation

tert-Butyl 3-cyano-4-oxo-pyrrolidine-1-carboxylate

  • MDL number MFCD01318456
  • Cas Number : 175463-32-8
  • Molecular Weight : 210.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/0404.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/0404.html
pictograms GHS05, GHS07
hstatements H302, H315, H318, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart