SY3H3249F16C Synthonix Corporation

tert-Butyl 3-hydroxy-3-methyl-piperidine-1-carboxylate

  • MDL number MFCD13194152
  • Cas Number : 1104083-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/0541.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/0541.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart