SY3H3249F16E Synthonix Corporation

TERT-BUTYL 6-OXOPIPERIDIN-3-YLCARBAMATE

  • MDL number MFCD13195280
  • Cas Number : 1245646-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/0707.html

Safety Information

pcodes P280 ,
MSDS https://synthonix.com/0707.html
hstatements H301 , H319 ,
Availability details for
Quantity
  • Availability
  • Add to Cart