SY3H3249F170 Synthonix Corporation

tert-Butyl 3-sulfanylpiperidine-1-carboxylate

  • MDL number MFCD10566038
  • Cas Number : 1017798-34-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/0775.html

Safety Information

MSDS https://synthonix.com/0775.html
hstatements H302 ,
Availability details for
Quantity
  • Availability
  • Add to Cart