SY3H3249F171 Synthonix Corporation

tert-Butyl 3-(2-ethoxy-2-oxo-ethyl)piperidine-1-carboxylate

  • MDL number MFCD08275144
  • Cas Number : 384830-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/0811.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/0811.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart