SY3H3249F176 Synthonix Corporation

tert-Butyl 9-oxo-4,8-diazaspiro[4.4]nonane-4-carboxylate

  • MDL number MFCD16039601
  • Cas Number : 1221818-45-6
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/0964.html

Safety Information

signalword Warning
pcodes P280
MSDS https://synthonix.com/0964.html
pictograms GHS06, GHS07
hstatements H301, H319
Availability details for
Quantity
  • Availability
  • Add to Cart