SY3H3249F177 Synthonix Corporation

tert-Butyl 11-oxo-5,10-diazaspiro[5.5]undecane-5-carboxylate

  • MDL number MFCD16039652
  • Cas Number : 1221818-91-2
  • Molecular Weight : 268.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/0981.html

Safety Information

signalword Warning
pcodes P280
MSDS https://synthonix.com/0981.html
pictograms GHS06, GHS07
hstatements H301, H319
Availability details for
Quantity
  • Availability
  • Add to Cart