SY3H3249F1A0 Synthonix Corporation

Butyl 2,2-difluorocyclopropanecarboxylate

  • MDL number No MDL found
  • Cas Number : 260352-79-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/13359.html

Safety Information

MSDS https://synthonix.com/13359.html
hstatements H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart