SY3H3249F1B5 Synthonix Corporation

3-(Benzyloxy)oxan-4-one

  • MDL number No MDL found
  • Cas Number : 1351450-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/15668.html

Safety Information

MSDS https://synthonix.com/15668.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart