SY3H3249F1B8 Synthonix Corporation

1-Benzyl-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD00085749
  • Cas Number : 5733-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/1595.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/1595.html
hstatements H315 , H317 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart